Learn More
BINDER™ Convenient Water Supply
Shop All BINDER Products
Click to view available options
Material:
Plastic
Unit Size:
Each
Description
- For versions with active humidification
- Flexible positioning
- Enables water to be supplied to multiple CB units simultaneously by water container
- Not pre-filled
- 10 L water container with magnetic holder
- Hose adapter with practical quick coupling and water stop function
Specifications
Specifications
| Capacity (English) | 2.6 gal. |
| Description | Convenient water supply - for versions with active humidification, Set consisting of 10 liter water container with magnetic holder, hose adapter with practical quick coupling and water stop function, flexible positioning, enables water to be supplied to multiple CB units simultaneously via a water container, not pre-filled. |
| For Use With (Equipment) | BINDER Chambers CB170, CB260 |
| Material | Plastic |
| Type | Water Supply |
| Includes | 10 L water container with magnetic holder, Hose adapter with practical quick coupling and water stop function |