Learn More
Micronova™ Controlled Environment Wipe
Product Code. 13107356 Shop All Micronova Products
Change view
Click to view available options
Dimensions (L x W):
457 x 457 mm
Unit Size:
Pack of 100
Product Code. Dimensions (L x W) Unit Size
13107356 457 x 457 mm Pack of 100
1 options
Product Code. 13107356 Supplier Micronova™ Supplier No. WP61818H
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Product Type Controlled Environment Wipe
Length (Metric) 457 mm
Width (Metric) 457 mm
Material Novapoly
Autoclavable Autoclavable
Dimensions (L x W) 457 x 457 mm
Packaging Quantity 1 Ea.
Ply 2-ply