missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Control DNA (from CEPH Individual 1347-02)

Product Code. 16375262 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
180 μL
Unit Size:
180µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16375262 180 μL 180µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16375262 Supplier Applied Biosystems™ Supplier No. 403062

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Contains control DNA in Tris buffer, pH 8.0, and EDTA.

The control DNA can be used with linkage mapping sets MD10, HD5, and LD20.

  • Functions as a control template to monitor PCR amplification
  • Serves as sizing reference for controlling gel-to-gel or capillary-to-capillary variation
  • Aids in the comparison of alleles sizes from different gels (allele binning)
  • Enables correlation with allele frequency data from external sources such as others databases and laboratories
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This product contains control DNA in Tris buffer, pH 8.0, and EDTA.
Format Tube
For Use With (Application) Sequencing
Includes Control Template(s)
No. of Reactions 150
Product Type Control DNA
Quantity 180 μL
Shipping Condition Dry Ice
Species Human

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.