missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Control DNA (from CEPH Individual 1347-02)
Shop All Applied Biosystems Products
Click to view available options
Quantity:
180 μL
Unit Size:
Each
Description
Includes
Contains control DNA in Tris buffer, pH 8.0, and EDTA.
- Functions as a control template to monitor PCR amplification
- Serves as sizing reference for controlling gel-to-gel or capillary-to-capillary variation
- Aids in the comparison of alleles sizes from different gels (allele binning)
- Enables correlation with allele frequency data from external sources such as others databases and laboratories
Specifications
Specifications
| Content And Storage | This product contains control DNA in Tris buffer, pH 8.0, and EDTA. |
| Format | Tube |
| For Use With (Application) | Sequencing |
| Includes | Control Template(s) |
| No. of Reactions | 150 |
| Product Type | Control DNA |
| Quantity | 180 μL |
| Shipping Condition | Dry Ice |
| Species | Human |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction