Description
- Lightweight, polyester, disposable
- Ideal for applying disinfectants and wet cleaning vertical surfaces, ceilings, and walls in controlled environments; reaching awkward corners and ceilings with a complete range of motion
- Absorbent foam carries solvents and disinfectants to the surface; polyester fabric offers excellent abrasion resistance and helps distribute the liquid uniformly on the surface being cleaned
- Heads are laser cut to seal edges, further limiting potential contamination in the mopping process
- Textured surface option is ideal for applications requiring additional scrubbing performance
- Contain no cellulose; compatible with a wide range of chemicals, detergents, disinfectants, and cleaners
Order Info
Aluminum handle with mop frame available separately (Cat. No. 19-038-555).
Specifications
Specifications
| Product Type | Mop Head |
| Sterility | Sterile |
| Autoclavable | Autoclavable |
| Size | Small |
| Disposable | Reusable |
| For Use With (Equipment) | 7.5 in. Regular Frame |
| For Use With (Application) | For applying disinfectants and wet cleaning ceilings and walls in controlled environments |
| Material | Knitted Polyester, Polyester Foam |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?