missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PROSAT™ PS-911 Presaturated Wipes

PROSAT™ wipes are an effective, efficient and convenient method of cleaning process tools and other equipment. PROSAT wipes are also ideal for wiping articles prior to pass through.

Brand:  Contec™ PS-911

Code : NEW

Additional Details : CAS Number : 67-63-0 Weight : 14.40000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II


Product Code. 15320871

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Presaturated with IPA and DI water blends in flat, resealable pouches individually lot-numbered for traceability
  • These specially designed pouches preserve cleanliness and solvent saturation levels
  • Solvent blend meets SEMI specifications for metals and contaminants
  • ISO Class 5 compatible
  • 30 wipes/pouch, 50 pouches/case
Specifications

Specifications

PROSAT™ Pre-saturated Wipes
ISO Class 5, SEMI for metals/contaminants
22.9 x 27.9 cm
Process tools and other equipment
70% IPA/ 30% DI Water
Polypropylene
Yes
Ideal for routine cleaning and wiping instruments and equipment
30 Wipes/Pouch, 50 Pouches/Cs.
Non-sterile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • CONTEC PROSAT MBPP WITH 70% IPA

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P370+P378-In case of fire: Use dry chemical, carbondioxide to extinguish.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains :

SDS
Documents

Documents

Certificates
Special Offers

Special Offers