Learn More
Contec™ EasyReach™ Cleaning Pads
Contec™'s EasyReach™ cleaning pads are very absorbent, which ensures even coverage of disinfectant or IPA.
Brand: Contec™ PSME0001
Additional Details : CAS Number : 7732-18-5 Weight : 3.20000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
- Single-use sealed polyester
- Provide a low level of particulates which is essential for use in product contact areas
- Very absorbent, ensuring good even coverage of disinfectant or IPA
- Easy grab edge on the cleaning pad ensures the pad fits tightly on the mop frame
- Sterile cleaning pads are presaturated with 70% IPA
Specifications
Presaturated Cleaning Pads (70% IPA/30%DI water) | |
Sterile | |
2.7 in. | |
8 Bags/Cs. | |
rigid, flexible film, half suits (CAI, CACI), Restricted Access Barrier Systems (RABS), Filling Lines, Laminar Air Flow Hoods (LAF), Bio-Safety Cabinets (BSC), Glove Boxes, Incubators |
7.5 in. | |
Not Autoclavable | |
Yes | |
8/Bag; 8 Bags/Cs. | |
Quilted Polyester |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.