Learn More
Buerkle™ Container Thread
Product Code. 31188862 Shop All Bürkle Products
Change view
Click to view available options
For Use With (Equipment):
Container Thread Bürkle Containers
Product Code. For Use With (Equipment)
31188862 Container Thread Bürkle Containers
1 options
Product Code. 31188862 Supplier Buerkle™ Supplier No. 34400034
  • ONLINE EXCLUSIVE PRICE
    £214.00
    £135.67 / Pack of 1
    Save £78.33
    36% Off
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
We will mount the container connector directly into the container for you. The thread connectors are possible anywhere. Please state the exact position on the container, if applicable.
  • Thread:G 3/4 Zoll
  • Version:External thread
  • Can be mounted on all Buerkle rectangular and round containers
TRUSTED_SUSTAINABILITY
Specifications
Type Tank Lid
For Use With (Equipment) Container Thread Bürkle Containers