missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Container, 75L, grey solid with hinged lid, 600mm x 400mm x 423mm
Brand: Manutan A022584
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Polypropylene containers with integral hinged lid that is held firmly closed by two clips on each side. All containers are European standard sizes.- Capacity: 75L
- Internal dimensions (H x W x D): 394mm x 555mm x 355mm