missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Container, 20L, grey, solid with hinged lid, 400mm x 300mm x 246mm
Description
Polypropylene containers with integral hinged lid that is held firmly closed by two clips on each side. All containers are European standard sizes.
- Capacity: 20L
- Internal dimensions (H x W x D): 220mm x 355mm x 255mm
Specifications
Specifications
| Capacity (Metric) | 20 L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?