missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Contactin 6 Polyclonal Antibody
Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at minus 20 °C for increased stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
The neural adhesion molecule Contactin-6, also known as NB-3, is a contactin/F3 subgroup member of immunoglobulin superfamily. It is expressed exclusively in the nervous system and mainly upregulated at the early postnatal stage during mouse brain development.
Specifications
Specifications
| Antigen | Contactin 6 |
| Applications | Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with no preservative |
| Gene | CNTN6 |
| Gene Accession No. | P97528 |
| Gene Alias | CNTN6; contactin 6; contactin-6; hNB-3; mNB-3; NB3; NB-3; neural adhesion molecule; neural recognition molecule NB-3 |
| Gene Symbols | CNTN6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?