missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Contactin 6 Polyclonal Antibody

Product Code. 11561922
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11561922 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11561922 Supplier Invitrogen™ Supplier No. PA118250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for increased stability, and at 4 °C with an appropriate antibacterial agent. Avoid repetitive freeze/thaw cycles. Glycerol (1:1) may be added for an additional stability.

The neural adhesion molecule Contactin-6, also known as NB-3, is a contactin/F3 subgroup member of immunoglobulin superfamily. It is expressed exclusively in the nervous system and mainly upregulated at the early postnatal stage during mouse brain development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Contactin 6
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with no preservative
Gene CNTN6
Gene Accession No. P97528, Q9JMB8, Q9UQ52
Gene Alias CNTN6; contactin 6; contactin-6; hNB-3; mNB-3; NB3; NB-3; neural adhesion molecule; neural recognition molecule NB-3
Gene Symbols CNTN6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues G Y P S P H Y R W K Q of mouse contactin-6 (aa 11-64).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27255, 27256, 53870
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.