missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Contactin 5 Recombinant Mouse Monoclonal Antibody (K29081_6F9)
GREENER_CHOICE

Product Code. 30830021
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830021 100 μL
30825865 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830021 Supplier Invitrogen™ Supplier No. MA564471
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: SGRSGRRHEL VIAWEPVSEE FQNGEGFGYI VAFRPNGTRG WKEKMVTSSE ASKFIYRDES VPPLTPFEVK VGVYNNKGDG PFSQIVVICS AEGEPSAAPT DVKATSVSVS EILVAWKHIK ESLGRPQGFE VGYWKDMEQE DTAETVKTRG NESFVILTGL EGNTLYHFTV RAYNGAGYGP PSSEVSATTK KSPPSQAPSN LRWEQQGSQV SLGWEPVIPL ANESEVVGYK VFYRQEGHSN SQVIETQKLQ AVVPLPDAGV YIIEVRAYSE GGDGTASSQI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%, Pig - 97%, Cynomolgus monkey - 99%.

The protein encoded by this gene is a member of the immunoglobulin superfamily, and contactin family, which mediate cell surface interactions during nervous system development. This protein is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Contactin 5
Applications Western Blot
Classification Recombinant Monoclonal
Clone K29081_6F9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene CNTN5
Gene Accession No. O94779
Gene Alias 6720426O10Rik; A830025P08Rik; CNTN5; contactin 5; contactin-5; Gm507; hNB-2; HNB-2s; MGC163491; Nb-2; neural adhesion molecule; neural recognition molecule NB-2
Gene Symbols CNTN5
Host Species Mouse
Immunogen Recombinant human CNTN5 (aa 781-1060).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53942
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.