missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Contactin 5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558814
This item is not returnable.
View return policy
Description
Immunogen sequence: KSLPGLSTSY AALLRIKKSS SSSLFGSKTR PRYSSPSLGT LSASSPSWLG AAQNYYSPIN LYHSSDAFKQ DESVDYGPVF VQEPDDII Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 84%.
The protein encoded by this gene is a member of the immunoglobulin superfamily, and contactin family, which mediate cell surface interactions during nervous system development. This protein is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| Contactin 5 | |
| Polyclonal | |
| Unconjugated | |
| CNTN5 | |
| 6720426O10Rik; A830025P08Rik; CNTN5; contactin 5; contactin-5; Gm507; hNB-2; HNB-2s; MGC163491; Nb-2; neural adhesion molecule; neural recognition molecule NB-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53942 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94779 | |
| CNTN5 | |
| Recombinant protein corresponding to Human Contactin 5. Recombinant protein control fragment (Product #RP-97378). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction