missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£342.00 - £589.00
Specifications
| Antigen | Contactin-5 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18417320
|
Novus Biologicals
NBP1-83243-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18079603
|
Novus Biologicals
NBP1-83243 |
0.1 mL |
£589.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Contactin-5 Polyclonal specifically detects Contactin-5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Contactin-5 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| contactin 5, contactin-5, hNB-2, HNB-2s, MGC163491, NB2, NB-2, neural adhesion molecule, Neural recognition molecule NB-2 | |
| CNTN5 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 53942 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:IVVICSAEGEPSAAPTDVKATSVSVSEILVAWKHIKESLGRPQGFEVGYWKDMEQEDTAETVKTRGNESFVILTGLEGNTLYHFTVRAYNGAGYG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title