missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin 4 Polyclonal Antibody

Brand: Thermo Scientific PA5113101
This item is not returnable.
View return policy
Description
Specifications
| Contactin 4 | |
| Polyclonal | |
| Unconjugated | |
| CNTN4 | |
| 9630050B05; Axcam; axonal-associated cell adhesion molecule; Big-2; brain-derived immunoglobulin superfamily molecule; brain-derived immunoglobulin superfamily protein 2; CNTN4; CNTN4A; contactin 4; contactin-4; Kiaa3024; MGC33615; neural cell adhesion protein BIG-2; SCA16 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 152330 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IWV2 | |
| CNTN4 | |
| Recombinant Human Contactin-4 protein (777-1026AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction