missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-4 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98305AF405
This item is not returnable.
View return policy
Description
Contactin-4 Polyclonal specifically detects Contactin-4 in Mouse samples. It is validated for ELISA.
Specifications
| Contactin-4 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 152330 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 405 | |
| AXCAM, axonal-associated cell adhesion molecule, BIG-2Brain-derived immunoglobulin superfamily protein 2, contactin 4, contactin-4, MGC33615, neural cell adhesion protein BIG-2 | |
| Produced in rabbits immunized with purified, recombinant mouse Contactin-4 (Met1-Gly999). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu