missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-3 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£214.00
Specifications
| Antigen | Contactin-3 |
|---|---|
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Contactin-3 Polyclonal antibody specifically detects Contactin-3 in Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
| Contactin-3 | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| contactin 3 (plasmacytoma associated) | |
| Purified, recombinant Rat Contactin-3 (Q62682; Met1-Gly1001) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2μm filtered solution in PBS without Preservative | |
| 5067 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title