missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Contactin 3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149024
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Contactin 3 is a type of contactin that mediate cell surface interactions during nervous system development. Has some neurite outgrowth-promoting activity.
Specifications
| Contactin 3 | |
| Polyclonal | |
| Unconjugated | |
| CNTN3 | |
| BIG-1; Brain-derived immunoglobulin superfamily protein 1; Cntn3; contactin 3; contactin 3 (plasmacytoma associated); contactin-3; KIAA1496; Pang; PCS; Plasmacytoma-associated neuronal glycoprotein; Plasmacytoma-associated neuronal glycoprotein (neural cell adhesion molecule BIG-1) | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 18488, 5067, 54279 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q07409, Q62682, Q9P232 | |
| CNTN3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Contactin-3, Glu20-Thr1003. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction