missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Contactin 3 Monoclonal Antibody (524518)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561698

Product Code. 30534648

  • £374.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

Contactin 3 is a type of contactin that mediate cell surface interactions during nervous system development. Has some neurite outgrowth-promoting activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contactin 3
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q07409
CNTN3
Chinese hamster ovary cell line CHO-derived recombinant mouse Contactin-3, Glu20-Thr1003.
100 μg
Primary
Mouse
Antibody
IgG2a
Flow Cytometry, Western Blot
524518
Unconjugated
CNTN3
BIG-1; Brain-derived immunoglobulin superfamily protein 1; Cntn3; contactin 3; contactin 3 (plasmacytoma associated); contactin-3; KIAA1496; Pang; PCS; Plasmacytoma-associated neuronal glycoprotein; Plasmacytoma-associated neuronal glycoprotein (neural cell adhesion molecule BIG-1)
Rat
Protein A/G
RUO
18488
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.