missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-3 Antibody (524518), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB6644APCCY7
This item is not returnable.
View return policy
Description
Contactin-3 Monoclonal antibody specifically detects Contactin-3 in Mouse samples. It is validated for Flow Cytometry
Specifications
| Contactin-3 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5067 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry | |
| 524518 | |
| Flow Cytometry | |
| contactin 3 (plasmacytoma associated) | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Contactin-3, Glu20-Thr1003, Accession # Q07409 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction