missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Contactin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578380
This item is not returnable.
View return policy
Description
Positive Control: mouse brain, rat brain Predicted Reactivity: Human (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Contactin 1 encodes a protein that is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| Contactin 1 | |
| Polyclonal | |
| Unconjugated | |
| Cntn1 | |
| AW495098; CNTN; CNTN1; contactin 1; contactin-1; F3; F3cam; glycoprotein gP135; GP135; MYPCN; neural cell surface protein F3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117258, 12805 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot | |
| 2.25 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P12960, Q63198 | |
| Cntn1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of mouse Contactin 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction