missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Contactin 1 Monoclonal Antibody (10B3B1D1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA529177
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CNTN1/Contactin-1.
Contactin 1 encodes a protein that is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| Contactin 1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q12860 | |
| Cntn1 | |
| Recombinant Human CNTN1 protein (Met1-Ser993). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA | |
| 10B3B1D1 | |
| Unconjugated | |
| Cntn1 | |
| AW495098; CNTN; CNTN1; contactin 1; contactin-1; F3; F3cam; glycoprotein gP135; GP135; MYPCN; neural cell surface protein F3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 1272 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction