Learn More
uvex Contact Ergo ENB 20C Safety Glove
Product Code. 17747609 Shop All Uvex Products
Click to view available options
Size:
7
8
9
Unit Size:
10 pairs
Product Code. 17747609
Brand: uvex 6015001
Only null left
Add to basket
Add to basket
The thick special NBR coating of the Uvex contact ergo is resistant to oil and grease.
  • Thick, hard-wearing cotton interlock safety glove
  • Palm and fingers with special NBR coating (nitrile rubber)
  • Good grip in damp, wet and oily areas
  • Good tactile feel
  • Ergonomic fit
  • High flexibility
  • Excellent wearer comfort with the high water vapor absorption of the cotton lining
  • Free of harmful substances in line with OEKO-Tex Standard 100
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 388:2016 (2 1 2 1 X)
Coating Material NBR
Color Orange, White
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Cotton
Product Type Mechanical Protection Gloves
Size 7