Learn More
Specac™ Constant Thickness Film Maker Kit I
Product Code. 15240178
Encompass_Preferred
Change view
Click to view available options
Type:
Constant Thickness Film Maker Kit I
Product Code. Type
15240178 Constant Thickness Film Maker Kit I
1 options
Product Code. 15240178 Supplier Specac™ Supplier No. GS15631
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This kit includes a film maker accessory GS15640 and Atlas Heated Platens GS15515 for the preparation of thin polymer films. This kit does not include a Manual Press.
Specifications
Type Constant Thickness Film Maker Kit I
Includes A film maker accessory GS15640 and Atlas Heated Platens GS15515 for the preparation of thin polymer films