Learn More
Fistreem International™ Constant Level Device
Product Code. 13186010
Click to view available options
For Use With (Equipment):
Calypso or Cyclon Ultrapure Water Still
Unit Size:
Each
Product Code. 13186010
Brand: Fistreem International™ WSC707.CLJ.9
Only null left
Add to basket
Add to basket
Replacement constant level device for the Calypso or Cyclon Ultrapure Water Still. Fistreem International™ Constant Level Device allows cleaning solution to be simply introduced into the boiler with no dismantling.
  • Borosilicate glass construction.
  • Suitable for Calypso or Cyclon Ultrapure Water Still.
Specifications
Weight (Metric) 5 kg
For Use With (Equipment) Calypso or Cyclon Ultrapure Water Still
Type Constant Level Device