missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Constans Safety Shoes

Product Code. 10173974
Change view
Click to view available options
Size:
40
46
Unit Size:
1 pair
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10173974 46 1 pair
15287038 40 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10173974 Supplier Emma Safety Footwear Supplier No. MM13207046
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Developed for semiorthopedic adjustments requiring customized in-soles.

  • Full grain leather upper construction
  • Nappa leather lining
  • With sole plate for wet work
  • Woven zero penetration in-sole
  • Antibacterial inlay sole
  • Vibram™ Nitril / HRO outsole

Specifications

Color Black
For Use With (Application) semiorthopedic adjustments requiring customized in-soles
Liner Material Nappa leather
Material Leather
Size 46
Sole Features Woven zero-penetration insole, Heat-resistant outsole
Sole Material Vibram™ Nitrile
Product Type Constans Safety Shoes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.