missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Connexin 50 Monoclonal Antibody (C6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ 334300
This item is not returnable.
View return policy
Description
Connexin 50 Monoclonal Antibody for ELISA, ICC/IF, IP, Western Blot
This gene encodes a transmembrane connexin protein that is necessary for lens growth and maturation of lens fiber cells. The encoded protein is a component of gap junction channels and functions in a calcium and pH-dependent manner. Mutations in this gene have been associated with zonular pulverulent cataracts, nuclear progressive cataracts, and cataract-microcornea syndrome.
Specifications
| Connexin 50 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P28236, P48165, P55917, Q8K4Q9 | |
| GJA8 | |
| This antibody was prepared against urea/alkali stripped sheep lens membranes. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Sheep | |
| Antibody | |
| IgM κ |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| C6 | |
| Unconjugated | |
| GJA8 | |
| Aey5; alpha 8 connexin; CAE; CAE1; cell surface glycoprotein; Cnx50; connexin 50; connexin-49; Connexin-50; CTRCT1; cx49; CX50; CZP1; gap junction alpha 8; gap junction alpha-8 protein; gap junction membrane channel protein alpha 8; gap junction membrane channel protein alpha-8; gap junction protein alpha 8; gap junction protein alpha 8 50kDa; gap junction protein, alpha 8; gap junction protein, alpha 8, 50kDa; GJA8; Lens fiber protein MP70; lens intrinsic membrane protein MP70; lens opacity 10; Lop10; MP38; MP64; MP70 | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| 100170231, 14616, 2703, 29601 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction