missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 50/GJA8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-15748-100UL
This item is not returnable.
View return policy
Description
Connexin 50/GJA8 Polyclonal antibody specifically detects Connexin 50/GJA8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Connexin 50/GJA8 | |
| Polyclonal | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| alpha 8, 50kD (connexin 50), CAE, Connexin-50, Cx50, CZP1, gap junction alpha-8 protein, gap junction membrane channel protein alpha-8, gap junction protein, alpha 8, 50kDa, gap junction protein, alpha 8, 50kDa (connexin 50), lens intrinsic membrane protein MP70, MP70 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 234-433 of human Connexin 50/GJA8 (NP_005258.2). | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 2703 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction