missing translation for 'onlineSavingsMsg'
Learn More

Connexin 45/GJC1 Antibody (5B9.2), DyLight 594, Novus Biologicals™

Product Code. 18413347 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413347 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413347 Supplier Novus Biologicals Supplier No. NBP229754DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Connexin 45/GJC1 Monoclonal antibody specifically detects Connexin 45/GJC1 in Human, Rat, Rabbit samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 45/GJC1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 5B9.2
Conjugate DyLight 594
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias connexin 45, Connexin-45, CX45, DKFZp686P0738, Gap junction alpha-7 protein, gap junction gamma-1 protein, gap junction protein, alpha 7, 45kDa, gap junction protein, gamma 1, 45kDa, GJA7
Host Species Mouse
Immunogen The peptide sequence used for the immunogen was H-Gln-Ala-Tyr-Ser-His-Gln-Asn-Asn-Pro-His-Gly-Pro-Arg-Glu-Cys-OH conjugated to KLH, corresponding to amino acids 354-377 of human connexin 45.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10052
Target Species Human, Rat, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.