missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 45/GJC1 Antibody (5B9.2), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29754R
This item is not returnable.
View return policy
Description
Connexin 45/GJC1 Monoclonal antibody specifically detects Connexin 45/GJC1 in Human, Rat, Rabbit samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Connexin 45/GJC1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Rabbit | |
| Purified |
| Western Blot, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 5B9.2 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| connexin 45, Connexin-45, CX45, DKFZp686P0738, Gap junction alpha-7 protein, gap junction gamma-1 protein, gap junction protein, alpha 7, 45kDa, gap junction protein, gamma 1, 45kDa, GJA7 | |
| The peptide sequence used for the immunogen was H-Gln-Ala-Tyr-Ser-His-Gln-Asn-Asn-Pro-His-Gly-Pro-Arg-Glu-Cys-OH conjugated to KLH, corresponding to amino acids 354-377 of human connexin 45. | |
| 0.1 mL | |
| Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 10052 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction