missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 43/GJA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£160.00 - £389.00
Specifications
| Antigen | Connexin 43/GJA1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18269424
|
Novus Biologicals
NB100-81867-0.025ML |
0.025 mL |
£160.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18189879
|
Novus Biologicals
NB100-81867 |
0.1 mL |
£389.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Connexin 43/GJA1 Polyclonal specifically detects Connexin 43/GJA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Connexin 43/GJA1 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| P17302 | |
| 2697 | |
| IgG | |
| Affinity Purified | |
| 43 kDa |
| 1.0 mg/mL | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Mouse, Rat, Human | |
| connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJAL, HSS, ODD, ODDD, ODOD, SDTY3 | |
| GJA1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title