missing translation for 'onlineSavingsMsg'
Learn More

Connexin 43/GJA1 Antibody (105), DyLight 350, Novus Biologicals™

Product Code. 18319910 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319910 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319910 Supplier Novus Biologicals Supplier No. NBP289209UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Connexin 43/GJA1 Monoclonal specifically detects Connexin 43/GJA1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 43/GJA1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 105
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJAL, HSS, ODD, ODDD, ODOD, SDTY3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Connexin 43/GJA1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2697
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.