missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 40/GJA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£175.00 - £416.00
Specifications
| Antigen | Connexin 40/GJA5 |
|---|---|
| Concentration | LYOPH |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18288254
|
Novus Biologicals
NBP1-46568-0.025ML |
0.025 mL |
£175.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18282203
|
Novus Biologicals
NBP1-46568 |
0.1 mL |
£416.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Connexin 40/GJA5 Polyclonal specifically detects Connexin 40/GJA5 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Connexin 40/GJA5 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| 2702 | |
| IgG | |
| Unpurified | |
| Specific for Connexin-40. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| alpha 5, 40kD (connexin 40), connexin-40, Cx40, gap junction alpha-5 protein, gap junction protein, alpha 5, 40kDa, gap junction protein, alpha 5, 40kDa (connexin 40), MGC11185 | |
| GJA5 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title