missing translation for 'onlineSavingsMsg'
Learn More

Connexin 30/GJB6 Antibody (CL4540) [Alexa Fluor™ 488], R&D Systems™

Product Code. 30297103 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30297103 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30297103 Supplier Novus Biologicals Supplier No. NBP261141AF488
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Connexin 30/GJB6 Monoclonal antibody specifically detects Connexin 30/GJB6 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 30/GJB6
Applications Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL4540
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias connexin 30, connexin-30, Cx30, CX30gap junction protein, beta 6, DFNA3B, DFNB1B, ectodermal dysplasia 2, hidrotic (Clouston syndrome), ED2, EDH, gap junction beta-6 protein, gap junction protein, beta 6 (connexin 30), gap junction protein, beta 6, 30kDa, HEDDFNA3
Host Species Mouse
Immunogen This antibody was developed using a synthetic peptide derived from O95452, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10804
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.