missing translation for 'onlineSavingsMsg'
Learn More

Connexin 30.3/GJB4 Antibody (1E3-1C12), Novus Biologicals™

Product Code. 18418238 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418238 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418238 Supplier Novus Biologicals Supplier No. H00127534M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Connexin 30.3/GJB4 Monoclonal antibody specifically detects Connexin 30.3/GJB4 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 30.3/GJB4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1E3-1C12
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH34709
Gene Alias beta 4, connexin-30.3, Cx30.3, EKV, gap junction beta-4 protein, gap junction protein, beta 4 (connexin 30.3), gap junction protein, beta 4, 30.3kDa, MGC21116
Host Species Mouse
Immunogen GJB4 (AAH34709, 1 a.a. ∽ 266 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MNWAFLQGLLSGVNKYSTVLSRIWLSVVFIFRVLVYVVAAEEVWDDEQKDFVCNTKQPGCPNVCYDEFFPVSHVRLWALQLILVTCPSLLVVMHVAYREERERKHHLKHGPNAPSLYDNLSKKRGGLWWTYLLSLIFKAAVDAGFLYIFHRLYKDYDMPRVVACSVEPCPHTVDCYISRPTEKKVFTYFMVTTAAICILLNLSEVFYLVGKRCMEIFGPRHRRPRCRECLPDTCPPYVLSQGGHPEDGNSVLMKAGSAPVDAGGYP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127534
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.