missing translation for 'onlineSavingsMsg'
Learn More

Connexin 26/GJB2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications

£150.00 - £428.00

Specifications

Antigen Connexin 26/GJB2
Concentration 1 mg/mL
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18690558
View Documents
Novus Biologicals
NBP2-41304-0.025mg
0.025 mg
£150.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18043064
View Documents
Novus Biologicals
NBP2-41304
0.1 mg
£428.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Connexin 26/GJB2 Polyclonal specifically detects Connexin 26/GJB2 in Human, Rat, Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Connexin 26/GJB2
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Rat, Pig
beta 2, 26kD (connexin 26), Cx26, CX26DFNA3, DFNA3A, DFNB1, gap junction protein, beta 2, 26kDa, gap junction protein, beta 2, 26kDa (connexin 26), NSRD1
GJB2
IgG
Affinity Purified
26 kDa
1 mg/mL
Polyclonal
Rabbit
Neuroscience, Sensory Systems
PBS with 0.02% Sodium Azide
2706
Antibody was raised against a 16 amino acid peptide near the center of human GJB2. The immunogen is located within amino acids 80 - 130 of GJB2. Amino Acid Squence: HVAYRRHEKKRKFIKGE
Primary
GJB2 antibody is human specific.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.