missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Connexin 25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140048
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: YKLYDGFSVP YLIKCDLKPC PNTVDCFISK PTEKTIFI Highest antigen sequence indentity to the following orthologs: Mouse - 68%, Rat - 68%.
Connexins, such as GJB7, are involved in the formation of gap junctions, intercellular conduits that directly connect the cytoplasms of contacting cells. Each gap junction channel is formed by docking of 2 hemichannels, each of which contains 6 connexin subunits (Sohl et al., 2003 [PubMed 12881038]).[supplied by OMIM, Mar 2008].
Specifications
| Connexin 25 | |
| Polyclonal | |
| Unconjugated | |
| GJB7 | |
| bA136M9.1; connexin 25; connexin25; connexin-25; CX25; Gap junction beta-7 protein; gap junction protein beta 7; gap junction protein, beta 7, 25kDa; GJB7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 375519 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PEY0 | |
| GJB7 | |
| Recombinant protein corresponding to Human GJB7. Recombinant protein control fragment (Product #RP-108726). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction