missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SCAT™ connectors are available in the following versions:
- Conical connectors
- Y-connectors
- Tube connectors
- Capillary connectors
Specifications
Specifications
| Diameter (Metric) | 6.5 to 10 mm |
| Autoclavable | Autoclavable |
| Material | PTFE |
| For Use With | Tubing Connector |
| For Use With (Application) | HPLC/UHPLC Systems |
| Includes | Locknut |
| Color | White |
| Product Type | Compression Fitting |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?