missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Connector Cables for Automated Thermal Cycler (ATC)

Product Code. 15602436 Shop All Applied Biosystems Products
Click to view available options
Format:
1-m connector cable (flexible)
3-m connector cable (flexible)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15602436

Brand: Applied Biosystems™ A31482

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

1 meter connection cable for Automated Thermal Cycler (ATC)

Used to connect the Automated Thermal Cycler (ATC) block to the power box, this connector is available in a rigid 10-cm, flexible 1-m, or flexible 3-m cable format.

For customers seeking to automate their PCR workflow, the Applied Biosystems Automated Thermal Cycler (ATC) provides the reliability and performance you've come to expect from Applied Biosystems instruments. Small, easy-to-integrate, with a compatible format, our Automated Thermal Cycler enables hands-free PCR results.

In order to better align with lab design and demands, a longer or shorter cable may be utilized to optimize workflow.

Warranty

27 month warranty

TRUSTED_SUSTAINABILITY

Specifications

Format 1-m connector cable (flexible)
For Use With (Equipment) Automated Thermal Cycler (ATC)
Product Type Connector Cable
For Use With (Application) PCR
Max. Heating Rate (Thermal Block) 2.8°C/sec. (Block), 1.6°C/sec. (Sample)
Quantity 1 cable

For Research Use Only. Not for use in diagnostic procedures.

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.