Learn More
Manutan Connector for 7mm chain
Product Code. 12818558
Encompass_Preferred
Product Code. 12818558 Supplier Manutan Supplier No. A341465
Only null left
Add to basket
Add to basket
A range of high strength chain and fittings to suit a variety of lifting, lashing, suspending or security applications. Fittings - 'connector' for linking components and/or chain consists of 2 matched halves joined by a cross pin and tension bush. Other fittings are connected to chain using the Kuplex clevis system with load retaining pin. Test certificate supplied
  • Connector
  • Chain diameter: 7mm
  • Workload Limit: 1500