Learn More
WTW™ Connection Cable for Depth Probes
For use with WTW™ MPP IDS depth probes with waterproof plug head.
£120.00 - £174.00
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
15100173
|
WTW™
903854 |
15 m |
£174.00
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
15110173
|
WTW™
903853 |
10 m |
£152.00
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
13466768
|
WTW™
903852 |
6 m |
£133.00
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
15230396
|
WTW™
903851 |
3 m |
£120.00
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Description
- Also useful for plug head sensors
- Available in a range of lengths
- Supplied individually
Specifications
Connection Cable |