Learn More
BOLA Connection Bolts
Product Code. 10767522
Click to view available options
Includes:
Fingertight Nut, Single Ferrule
Hex-head Nut, Double Ferrule
Unit Size:
Each
Product Code. 10767522
Brand: BOLA F83310
Only null left
Add to basket
Add to basket
Connection Bolts Poly Ether Ether Ketone (PEEK)
  • Connection bolt and sealing cone made of PEEK, suitable for female thread UNF 10-32 G.
  • Exchangeable sealing cone available separately, knurled nut reusable.
  • Suitable for pressures up to max.
  • 280 bar
Specifications
Diameter (English) Outer 1.6 in.
Includes Fingertight Nut, Single Ferrule
Autoclavable Autoclavable