Learn More
Lenz Laborglasintrumente™ Connecting Spring for Ground Joints
Click to view available options
Size:
40 mm L
Unit Size:
Pack of 10
Description
- Stainless Steel
- Two rings
Specifications
Specifications
| Quantity | 10 Pk. |
| Material | Stainless Steel |
| Includes | Two Rings |
| For Use With (Application) | Connecting conical ground joints |
| Size | 40 mm L |