Learn More
HUBER USB Control Cable
Product Code. 15286537
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
15286537 1 Ea. Each
1 options
Product Code. 15286537 Supplier HUBER Supplier No. 54949
  • ONLINE EXCLUSIVE PRICE
    £22.95
    £22.00 / Pack of 1
    Save £0.95
    4% Off
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
USB control cable from: controller Pilot ONE, Mini-USB, to: PC / Notebook, USB Type A. For data recording, data communication and firmware updates.
Specifications
Product Type USB Control Cable
Quantity 1 Ea.