Learn More
Lenz Laborglasintrumente™ Conical Ground Joint Cone
Product Code. p-7161767
Click to view available options
Cone Size:
NS 10/19
NS 34/35
Unit Size:
Pack of 10
Product Code. 11953351
Brand: Lenz Laborglasintrumente™ 01100210
Only null left
Add to basket
Add to basket
Conical ground cones made of DURAN™ tubing
  • DIN 12249
  • 10 pack
Specifications
Quantity 10
Item Description Cone made from Boro 3.3
Cone Size NS 10/19
Thickness (Metric) Wall 1.5 mm
Diameter (Metric) Outer Tube 8 mm