missing translation for 'onlineSavingsMsg'
Learn More

Congo red, pure, C.I. 22120, pH and adsorption indicator, Fisher Chemical

Product Code. 10447060 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10447060 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10447060 Supplier Fisher Chemical Supplier No. C/7020/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 360°C
CAS 573-58-0
Molecular Formula C32H22N6Na2O6S2
MDL Number 4028
Formula Weight 696.65g/mol
Packaging Glass Jar
Color Brown/Red
pH 8 to 9.5
Quantity 25 g
Physical Form Powder
compliance-icons
Product Identifier
  • Congo red
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H350-May cause cancer.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P233-Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.