missing translation for 'onlineSavingsMsg'
Learn More

Congo Red, MP Biomedicals™

Product Code. p-93038 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g

Product Code. 11422092

Brand: MP Biomedicals 0215071105

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 573-58-0
Molecular Formula C32H22N6Na2O6S2
Molecular Weight (g/mol) 696.664
InChI Key IQFVPQOLBLOTPF-UHFFFAOYSA-L
Synonym Cotton Red, Benzo Congo Red
PubChem CID 11313
ChEBI CHEBI:34653
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Formula Weight 696.7
Color Red
Quantity 5 g
Physical Form Powder
Chemical Name or Material Congo red
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.