missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Congo Red, indicator grade

Product Code. 11435737
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435737 500 g 500g
11415737 25 g 25g
11425737 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435737 Supplier Thermo Scientific Chemicals Supplier No. B24310.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 573-58-0 | C32H22N6Na2O6S2 | 696.664 g/mol

Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

Solubility
Soluble in water 25 g/L.

Notes
Store away from oxidizing agents, acids, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 573-58-0
Molecular Formula C32H22N6Na2O6S2
Molecular Weight (g/mol) 696.664
MDL Number MFCD00004028
InChI Key IQFVPQOLBLOTPF-UHFFFAOYSA-L
Synonym C.I. 22120
PubChem CID 11313
ChEBI CHEBI:34653
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >360°C
Odor Odorless
Beilstein 3894858
Merck Index 14,2498
Solubility Information Soluble in water 25g/L.
Quantity 500 g
Formula Weight 696.67
Grade Indicator
Chemical Name or Material Congo Red

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.