missing translation for 'onlineSavingsMsg'
Learn More

Congo Red (90%), TRC

Product Code. 30133730
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133730 10 g 10g
17928459 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133730 Supplier Toronto Research Chemicals Supplier No. TRCC68505010G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 573-58-0
Molecular Formula C32 H22 N6 O6 S2 . 2 Na
Molecular Weight (g/mol) 696.66
Synonym 1-Naphthalenesulfonic acid, 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis[4-amino-, disodium salt (9CI), C.I. Direct Red 28, disodium salt (8CI), Atlantic Congo Red, Atul Congo Red, Azocard Red Congo, Benzo Congo Red, Brasilamina Congo 4B, C.I. 22120, C.I. Direct Red 28, Congo Red, Congo red, Congo Red 4B, Congo Red 4BS, Congo Red 4BX, Congo Red CR, Congo red disodium salt, Congo Red H, Congo Red K, Congo Red L, Congo Red M, Congo Red N, Congo Red R, Congo Red RS, Congo Red sodium salt, Congo Red TS, Congo Red W, Congo Red WS, Cotton Red 4BC, Cotton Red 5B, Cotton Red L, Diacotton Congo Red, Direct Congo Red, Direct Red 28, Direct Red C, Direct Red DC-CF, Direct Red K, Direct Red R, Direct Scarlet 4B, Erie Congo 4B, Haemomedical, Haemonorm, Hemorrhagyl, Hispamin Congo 4B, Kayaku Congo Red, MeSH ID: D003224, Mitsui Congo Red, Peeramine Congo Red, Red K (dye), Solucongo, Sugai Congo Red, Tertrodirect Red C, Trisulfon Congo Red, Vondacel Red CL, 1-Naphthalenesulfonic acid, 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(2,1-diazenediyl)]bis[4-amino-, sodium salt (1:2) (ACI)
IUPAC Name disodium;4-amino-3-[(E)-[4-[4-[(E)-(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
SMILES [Na+].[Na+].Nc1c(cc(c2ccccc12)S(=O)(=O)[O-])N=Nc3ccc(cc3)c4ccc(cc4)N=Nc5cc(c6ccccc6c5N)S(=O)(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C32H24N6O6S2.2Na/c33-31-25-7-3-1-5-23(25)29(45(39,40)41)17-27(31)37-35-21-13-9-19(10-14-21)20-11-15-22(16-12-20)36-38-28-18-30(46(42,43)44)24-6-2-4-8-26(24)32(28)34;;/h1-18H,33-34H2,(H,39,40,41)(H,42,43,44);;/q;2*+1/p-2
Formula Weight 696.0838
Chemical Name or Material Direct Red 28
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.